missing translation for 'onlineSavingsMsg'
Learn More

p53 Antibody (PAb 240) - BSA Free, Novus Biologicals™

Product Code. 18700214 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18700214 0.025 mL 25µL
18797733 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18700214 Supplier Novus Biologicals Supplier No. NB200103SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 31 publication

p53 Monoclonal specifically detects p53 in Human, Mouse, Rat, Yeast, Xenopus (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone PAb 240
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:250-1:500, Immunocytochemistry/Immunofluorescence 5 ug/ml. Use reported in scientific literature (PMID 1394225), Immunoprecipitation 10ug/mg, Immunohistochemistry-Paraffin 1:250-1:500, Immunohistochemistry-Frozen reported in multiple pieces of scientific literature, Flow (Intracellular), CyTOF-ready
Gene Alias Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53
Gene Symbols TP53
Host Species Mouse
Immunogen Gel-purified p53-beta-galactosidase fusion protein containing murine p53 from aa 14-389 (derived from pSV53C cDNA clone).
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Hypoxia, p53 Pathway, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7157
Test Specificity This monoclonal recognizes both mutant forms and wild-type human p53 under denaturing conditions. It does not recognize nondenatured wild type p53. In nondenaturing conditions (IP) it only recognizes mutant p53.
Target Species Human, Mouse, Rat, Yeast, Xenopus (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.