missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P4HA3 Antibody (OTI9A2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44719
This item is not returnable.
View return policy
Description
P4HA3 Monoclonal antibody specifically detects P4HA3 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry
Specifications
| P4HA3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry | |
| OTI9A2 | |
| Immunohistochemistry 1:1000 | |
| 2-oxoglutarate-4-dioxygenase subunit alpha-3, 4-PH alpha-3, alpha polypeptide III, collagen prolyl 4-hydroxylase alpha(III), C-P4H alpha III, C-P4Halpha(III), EC 1.14.11.2, Procollagen-proline, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), prolyl 4-hydroxylase subunit alpha-3, prolyl 4-hydroxylase, alpha polypeptide III | |
| Full length human recombiant protein of human P4HA3 (NP_878907) produced in E.coli. | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 283208 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction