Learn More
P4HA2 Antibody, PerCP, R&D Systems™
Product Code. 18611786 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18611786 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18611786 Supplier Novus Biologicals Supplier No. NBP299486PCP
 Restricted Item
Estimated Shipment: 14-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
P4HA2 Polyclonal specifically detects P4HA2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mL
Regulatory Status RUO
Product Type Antibody
Product Line Novus