missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
p41 Flagellin Monoclonal antibody specifically detects p41 Flagellin in Bacteria samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | p41 Flagellin |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 6802 |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 41 kDa antigen, fla, Flagellar filament 41 kDa core protein, Flagellin, p41 |
| Host Species | Mouse |
| Immunogen | A crude sonicate of Borrelia spirochetes |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?