missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p40 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 326.00€
Specifications
| Antigen | p40 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18679645
|
Novus Biologicals
NBP2-99500-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18601496
|
Novus Biologicals
NBP2-99500-50ul |
50 μg |
216.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p40 Polyclonal antibody specifically detects p40 in Human samples. It is validated for Western BlotSpecifications
| p40 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 10244 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 40 kDa Rab9 effector protein, bA65N13.1, p40, Rab9 effector p40, Rab9 effector protein with kelch motifs, RAB9P40DKFZp686P1077, RABEPK | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human p40. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title