missing translation for 'onlineSavingsMsg'
Learn More

P2Y8 Antibody (1G5A11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

203.00€ - 552.00€

Specifications

Antigen P2Y8
Clone 1G5A11
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18282394
View Documents
Novus Biologicals
NBP2-61761
0.1 mL
552.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18658058
View Documents
Novus Biologicals
NBP2-61761-0.025ml
0.025 mL
203.00€
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

P2Y8 Monoclonal specifically detects P2Y8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

P2Y8
1 mg/mL
Monoclonal
Purified
Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers
286530
Purified recombinant fragment of human P2RY8 (AA: extra mix) expressed in E. Coli.
Primary
40.6 kDa
1G5A11
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000
Unconjugated
Mouse
G-protein coupled purinergic receptor P2Y8, P2Y purinoceptor 8, P2Y8MGC50878, purinergic receptor P2Y, G-protein coupled, 8
P2RY8
IgG2a
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.