missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y4/P2RY4 Antibody (3B4H6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61760-0.025ml
This item is not returnable.
View return policy
Description
P2Y4/P2RY4 Monoclonal specifically detects P2Y4/P2RY4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| P2Y4/P2RY4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:200, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| P2P, P2Y4P2Y purinoceptor 4, pyrimidinergic receptor P2Y, G-protein coupled, 4, UNRNRUUridine nucleotide receptor | |
| Mouse | |
| 41 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 3B4H6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| P2RY4 | |
| Purified recombinant fragment of human P2RY4 (AA: extra mix) expressed in E. Coli. | |
| Protein G purified | |
| GPCR | |
| 5030 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction