Learn More
Invitrogen™ P2Y10 Monoclonal Antibody (639329)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560852
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
P2Y10 belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. Two alternatively spliced transcript variants encoding the same protein isoform have been found for this gene. P2Y10 has been reported to be expressed in lymphoid cells and in monocytes derived from dendritic cells.
Specifications
| P2Y10 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O00398 | |
| P2RY10 | |
| NS0 mouse myeloma cell line transfected with human P2RY10. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 639329 | |
| Unconjugated | |
| P2RY10 | |
| 5830408N17Rik; G-protein coupled purinergic receptor P2Y10; LYPSR2; P2RY10; p2Y purinoceptor 10; P2Y10; P2Y-like receptor; purinergic receptor P2Y, G-protein coupled 10; purinergic receptor P2Y, G-protein coupled, 10; purinergic receptor P2Y10; putative P2Y purinoceptor 10; RGD1561912; RP3-333E23.1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 27334 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |