missing translation for 'onlineSavingsMsg'
Learn More

P2X7/P2RX7 Antibody (Hano43), PE/Cy5.5, Novus Biologicals™

Product Code. 18293611 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18293611 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18293611 Supplier Novus Biologicals Supplier No. NBP140894PECY55

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

P2X7/P2RX7 Monoclonal specifically detects P2X7/P2RX7 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2X7/P2RX7
Applications Flow Cytometry
Classification Monoclonal
Clone Hano43
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A
Gene Symbols P2RX7
Host Species Rat
Immunogen A P2X7-expression construct and a final boost with P2X7-transfected HEK cells.
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5027.0
Test Specificity This antibody recognises the P2X purinoceptor 7, also known as P2X7. The P2X7 ATP receptor has a distinctive long C-terminal tail with multiple potential protein and lipid interaction motifs and is highly polymorphic. It is a cation selective ion channel that opens up on binding of extracellular ATP. Sustained activation by extracellular ATP results in the formation of a reversible pore in the plasma membrane that is permeable to hydrophilic solutes of up to 900 Da. Once a pore is opened massive upset of cytoplasmic ion homeostasis occurs and the pore stays open as long as it is bound by ATP. Should ATP stimulation continue the cell will become irreversibly damaged and die. P2X7 plays a key role in the maturation and release of IL-1 and other IL-1 family members during inflammation. As such, P2X7 blockers might be useful as anti-inflammatory agents.
Target Species Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.