missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
P2X1/P2RX1 Monoclonal antibody specifically detects P2X1/P2RX1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
Specifications
| Antigen | P2X1/P2RX1 |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 551820 |
| Conjugate | DyLight 594 |
| Dilution | Flow Cytometry, CyTOF-ready |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ATP receptor, P2X purinoceptor 1, P2X receptor, subunit 1, P2X1P2X1 receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 1 |
| Host Species | Mouse |
| Immunogen | Human embryonic kidney cell, HEK293-derived transfected with human P2X1/P2RX1, Accession # P51575 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?