missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P16-INK4A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | P16-INK4A |
|---|---|
| Concentration | 0.37 mg/mL |
| Applications | Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16847563
|
Proteintech
10883-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16837563
|
Proteintech
10883-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
10883-1-AP reacts with p16 specifically. P16 plays an essential role in regulating the cell cycle, and mutations in p16 increase the risk of developing various cancers, including melanoma.
Kaiso is a novel member of the BTB/POZ (Broad complex, Tramtrak, Bric a brac/Pox virus and zinc finger) zinc finger superfamily of transcription factors. Yeast 2-hybrid studies have shown that Kaiso interacts with catenin p120ctn. This interaction involves the Arm repeats of 1-7 of the p120ctn Armadillo domain and the carboxyl-terminal third of Kaiso. The significance of the Kaiso:p120 ctn interaction is still unknown.Specifications
| P16-INK4A | |
| Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8N726 | |
| 1029 | |
| P16-INK4A Fusion Protein Ag1328 | |
| Primary | |
| -20°C | |
| CDKN2A |
| 0.37 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ARF, ARF,CDKN2A, CDK4 inhibitor p16 INK4, CDK4 inhibitor p16INK4, CDK4I, CDKN2, CDKN2A, CMM2, INK4, INK4a, MLM, MTS 1, MTS1, Multiple tumor suppressor 1, P16, p16 INK4, p16 INK4a, p16INK4, p16INK4a, TP16 | |
| CDKN2A | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title