missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p150 glued Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55182-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to DCTN1.
This gene encodes the largest subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. Dynactin is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit interacts with dynein intermediate chain by its domains directly binding to dynein and binds to microtubules via a highly conserved glycine-rich cytoskeleton-associated protein (CAP-Gly) domain in its N-terminus. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms. Mutations in this gene cause distal hereditary motor neuronopathy type VIIB (HMN7B) which is also known as distal spinal and bulbar muscular atrophy (dSBMA).Specifications
| p150 glued | |
| Polyclonal | |
| Unconjugated | |
| DCTN1 | |
| DAP 150, DCTN1, DP 150, Dynactin subunit 1, HMN7B, P135, p150 glued | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13191, 1639, 29167, 483106 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08788, P28023, Q14203 | |
| DCTN1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction