missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ P-Selectin/CD62P Antibody (inclacumab), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28137-50ug
This item is not returnable.
View return policy
Description
P-Selectin/CD62P Monoclonal antibody specifically detects P-Selectin/CD62P in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| P-Selectin/CD62P | |
| Monoclonal | |
| Unconjugated | |
| Q14242 | |
| Human | |
| Protein A purified | |
| RUO | |
| 6403 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| inclacumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
| P-Selectin/ CD62p | |
| 50 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction