missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxytetracycline 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Oxytetracycline |
| CAS | 79-57-2 |
| Analyte or Component Names | Oxytetracycline |
| Shipping Condition | -18°C |
| Molecular Formula | C22 H24 N2 O9 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Oxytetracycline, (4S,4aR,5S,5aR,6S,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo- (7CI,8CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-, [4S-(4α,4aα,5α,5aα,6β,12aα)]-, 5-Hydroxytetracycline, Abbocin, Adamycin, Antibiotic TM 25, Berkmycen, Biostat PA, Clinimycin, Dabicycline, Erfamycin, Fanterrin, Geomycin, Geomycin (Streptomyces vimosus), Geotilin, Imperacin, LA 200, Lenocycline, Liquamycin, Liquamycin LA 200, Macocyn, Mycoshield TMQTHC 20, NSC 9169, OTC, OTC (antibiotic), OXTC, Oksisyklin, Oxatets, Oxiter 200, Oxitetracyclin, Oxy-Kesso-Tetra, Oxycline, Oxydon, Oxyject LA, Oxylim, Oxymed 50, Oxymycin, Oxymykoin, Oxypam, Oxysentin 100, Oxyterracin, Oxyterracine, Oxyterracyne, Oxytetracyclin, Oxytetracycline, Oxytetracycline amphoteric, Oxytetral, Proteroxyna, Riomitsin, Ryomycin, Solkaciclina, Stecsolin, Stevacin, Stevasin, TM 50, Tarocyn, Tarosin, Teravit, Terrafungine, Terralon LA, Terramycin, Terramycin Q 50, Tetran, Unimycin, Ursocyclin, Ursocycline, Vendarcin |
| InChI Formula | InChI=1S/C22H24N2O9/c1-21(32)7-5-4-6-8(25)9(7)15(26)10-12(21)17(28)13-14(24(2)3)16(27)11(20(23)31)19(30)22(13,33)18(10)29/h4-6,12-14,17,25,27-29,32-33H,1-3H3,(H2,23,31)/t12-,13-,14+,17+,21-,22+/m1/s1 |
| SMILES | CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)cccc4[C@@]3(C)O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction