Learn More
Oxyphenonium Bromide, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Oxyphenonium Bromide |
| CAS | 50-10-2 |
| Analyte or Component Names | Oxyphenonium Bromide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H34 N O3 . Br |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethanaminium, 2-[(2-cyclohexyl-2-hydroxy-2-phenylacetyl)oxy]-N,N-diethyl-N-methyl-, bromide (1:1), Ethanaminium, 2-[(cyclohexylhydroxyphenylacetyl)oxy]-N,N-diethyl-N-methyl-, bromide (9CI), Antrenil, Antrenyl, Antrenyl Duplex, Ba-5473, C 5473, Oxifenon, Oxyfenon, Oxyphenon, Oxyphenonium bromide, Spasmodin, Spasmophen, dl-Oxyphenonium bromide, Ammonium, diethyl(2-hydroxyethyl)methyl-, bromide, alpha-phenylcyclohexaneglycolate (8CI), Diethyl(2-hydroxyethyl)methylammonium bromide alpha-phenylcyclohexaneglycolate (6CI), Cyclohexaneglycolic acid, alpha-phenyl-, ester with diethyl(2-hydroxyethyl)methylammonium bromide (8CI), (+/-)-Oxyphenonium bromide |
| InChI Formula | InChI=1S/C21H34NO3.BrH/c1-4-22(3,5-2)16-17-25-20(23)21(24,18-12-8-6-9-13-18)19-14-10-7-11-15-19;/h6,8-9,12-13,19,24H,4-5,7,10-11,14-17H2,1-3H3;1H/q+1;/p-1 |
| Solution Type | Neat |
| SMILES | [Br-].CC[N+](C)(CC)CCOC(=O)C(O)(C1CCCCC1)c2ccccc2 |
| Show More |