Learn More
Thermo Scientific Chemicals Oxymetazoline hydrochloride
Product Code. 16534384 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 16534384
Brand: Thermo Scientific Chemicals 463390250
Request Bulk or Custom Format
CAS: 2315-02-8 | C16H25ClN2O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2315-02-8
Molecular Formula C16H25ClN2O
Specifications
Product Type Oxymetazoline hydrochloride
Formula Weight 296.84
Quantity 25 g