accessibility menu, dialog, popup

UserName

CAS RN 2315-02-8

HClCH3OHCH3NHNH3CCH3H3C

CAS RN 2315-02-8

IUPAC Name: hydrogen 6-tert-butyl-3-[(4,5-dihydro-1H-imidazol-2-yl)methyl]-2,4-dimethylphenol chloride
Synonyms: oxymetazoline hydrochloride
Molecular Weight (g/mol): 296.84
Molecular Formula: C16H25ClN2O
InChi Key: BEEDODBODQVSIM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC(=C(O)C(C)=C1CC1=NCCN1)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results