missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXSM Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97290-50ul
This item is not returnable.
View return policy
Description
OXSM Polyclonal antibody specifically detects OXSM in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| OXSM | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| 3-ketoacyl-acyl carrier protein synthase, 3-oxoacyl-[acyl-carrier-protein] synthase, mitochondrial, 3-oxoacyl-ACP synthase, mitochondrial, acyl-malonyl acyl carrier protein-condensing enzyme, beta-ketoacyl synthase, Beta-ketoacyl-ACP synthase, EC 2.3.1, EC 2.3.1.41, FASN2D, FLJ20604, KASI, KS3-oxoacyl-acyl-carrier-protein synthase, type II mitochondrial beta-ketoacyl synthase | |
| Produced in rabbits immunized with E. coli-derived Human OXSM fragment. | |
| 50 μg | |
| metabolism | |
| 54995 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction