Learn More
Oxprenolol Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Oxprenolol Hydrochloride 1.0 mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Oxprenolol hydrochloride |
| CAS | 6452-73-9 |
| Analyte or Component Names | Oxprenolol hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H23 N O3 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propen-1-yloxy)phenoxy]-, hydrochloride (1:1), 2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propenyloxy)phenoxy]-, hydrochloride (9 CI), 2-Propanol, 1-[o-(allyloxy)phenoxy]-3-(isopropylamino)-, hydrochloride (7 CI,8 CI), 1-(2-Allyloxyphenoxy)-2-hydroxy-3-isopropylaminopropane hydrochloride, 1-(2'-Allyloxyphenoxy)-2-hydroxy-3-isopropylaminopropane hydrochloride, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba, CIBA 39089 Ba |
| InChI Formula | InChI=1 S/C15H23NO3.ClH/c1-4-9-18-14-7-5-6-8-15(14)19-11-13(17)10-16-12(2)3;/h4-8,12-13,16-17 H,1,9-11H2,2-3H3;1 H |
| SMILES | Cl.CC(C)NCC(O)COc1ccccc1OCC=C |
| IUPAC Name | 1-(propan-2-ylamino)-3-(2-prop-2-enoxyphenoxy)propan-2-ol;hydrochloride |
| Show More |