Learn More
Oxomemazine, LoGiCal
Product Code. 30457567
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457567 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30457567 Supplier LoGiCal Supplier No. LGCFOR0957.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Oxomemazine
CAS 3689-50-7
Analyte or Component Names Oxomemazine
Shipping Condition Room Temperature
Molecular Formula C18 H22 N2 O2 S
Certifications/Compliance ISO 9001
Synonym Trimeprazine Sulphone, Oxomemazine
InChI Formula InChI=1S/C18H22N2O2S/c1-14(12-19(2)3)13-20-15-8-4-6-10-17(15)23(21,22)18-11-7-5-9-16(18)20/h4-11,14H,12-13H2,1-3H3
Solution Type Neat
SMILES CC(CN(C)C)CN1c2ccccc2S(=O)(=O)c3ccccc13
IUPAC Name 3-(5,5-dioxophenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine
Industry Type Clinical Testing
Molecular Weight (g/mol) 330.44
Quantity 10 mg
Formula Weight 330.1402 g/mol