Learn More
Thermo Scientific™ Oxoid™ MRSV Selective Supplement
missing translation for 'orderingAttributeHoverText'
Quantity:
10/Pk.
missing translation for 'unitSize'
Pack of 10
Description
Oxoid MRSV Selective Supplement is used for the detection of Salmonella spp. from food and environmental samples.
- Add to MSRV Medium Base, Part No. CM0910B and Lysine Iron Agar, Modified (MLIA), Part No. CM0886B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | MSRV Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |