Learn More
Thermo Scientific™ Oxoid™ Modified Brucella Selective Supplement
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Description
Oxoid Modified Brucella Selective Supplement is for the cultivation and isolation of Brucella spp.
- Alternative supplement containing natamycin as a replacement for cycloheximide
- Add to Blood Agar Base No. 2, Part No. CM0271B or Brucella Medium Base, Part No. CM0169B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Modified Brucella Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Brucella species |