missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Listeria Selective Supplement
The Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
422.00€
Specifications
Species | Listeria |
---|---|
Description | Listeria Selective Supplement, Oxford |
Form | Freeze-dried |
Product Type | Selective Supplement |
Quantity | 10/Pk. |
Description
Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
Add to Listeria Selective Agar Base, Part No. CM0856BSpecifications
Listeria | |
Freeze-dried | |
10/Pk. |
Listeria Selective Supplement, Oxford | |
Selective Supplement |