Learn More
Thermo Scientific™ Oxoid™ Listeria Selective Supplement
The Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
Brand: Thermo Scientific™ SR0140E
Additional Details : CAS : 1264-72-8 Weight : 0.17500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : II
Description
Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
Add to Listeria Selective Agar Base, Part No. CM0856BSpecifications
Listeria | |
Freeze-dried | |
10/Pk. |
Listeria Selective Supplement, Oxford | |
Selective Supplement | |
Each vial for 500mL medium |
Safety and Handling
- LISTERIA Selective Supplement Oxford Formulation
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H300-Fatal if swallowed.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H360D-May damage the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Cycloheximide, Colistin, sulfate (salt),Acriflavin Neutral