Learn More
Thermo Scientific™ Oxoid™ C-T Supplement
Click to view available options
Yield:
Each vial for 2L medium
Each vial for 500mL medium
Unit Size:
Each
Pack of 10
Beskrivelse
Oxoid C-T Supplement is a chromogenic medium for the identification of Escherichia coli O157:H7.
Add to Sorbitol MacConkey Agar, Part No. CM0813B or to Sorbitol MacConkey Agar with BCIG, Part No. CM0981B.
Ordering Information:
Each vial supplements 2L of medium.
Tekniske data
Tekniske data
| Description | C-T Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion