Learn More
Thermo Scientific™ Oxoid™ Broad Spectrum Mixture Beta Lactamase
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Description
Oxoid Beta Lactamase, Broad Spectrum Mixture is a mixture of beta-lactamase enzymes from Bacillus cereus for the inactivation of beta-lactam antibiotic preparations present in specimens.
Specifications
Specifications
| Description | Beta Lactamase, Broad Spectrum Mixture |
| Form | Freeze-dried |
| Product Type | Beta Lactamase Mixture |
| Quantity | 10/Pk. |