Learn More
Thermo Scientific™ Brilliance Candida Selective Supplement
Product Code. 13285409
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13285409 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13285409 Supplier Thermo Scientific™ Supplier No. SR0231E
 Restricted Item
Estimated Shipment: 30-10-2026
Brilliance Candida is a chromogenic medium for the isolation and differentiation of Candida species.
  • Add to to Brilliance Candida Agar Base, Part No. CM1002A and CM1002B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Description Brilliance Candida Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium