missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ OXER1/5-oxo-ETE GPCR Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Specifications
Specifications
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1 M Urea, pH 7.4. |
| Gene ID (Entrez) | 165140 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 23 kDa |
| Quantity | 0.1 mL |
| Storage Requirements | Store at -20°C. Avoid freeze/thaw cycles. |
| Gene Alias | 5-oxo-ETE acid G-protein-coupled receptor 1,5-oxo-ETE G-protein coupled receptor, GPCR, GPR170G-protein coupled receptor R527, G-protein coupled receptor 170, oxoeicosanoid (OXE) receptor 1, oxoeicosanoid receptor 1, TG1019G-protein coupled receptor TG1019 |
| Gene Symbol | OXER1 |
| Inhibitor Properties | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-49006. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mLFor current lot information, including availability, please contact our technical support team click nb-technical@bio-techne.com For further blocking peptide related information and a protocol, click here . |
| Research Category | GPCR |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction