Learn More
OVOL1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93367-0.02ml
This item is not returnable.
View return policy
Description
OVOL1 Polyclonal antibody specifically detects OVOL1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| OVOL1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| HOVO1, ovo (Drosophila) homolog-like 1, ovo homolog-like 1, ovo-like 1(Drosophila), putative transcription factor Ovo-like 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human OVOL1 (NP_004552.2). MPRAFLVKKPCVSTCKRNWSELPDEERGEIYVPVSLGFCPPQPYREPEPSVAEPPSCPLALNMSLRDSSYSMAPGPCVVAQLPSEDMGHLTDPQSRDHGFLRTKMKVTLGDSPSGDLFTC | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5017 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |