missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Ovalbumin (Egg White)

Product Code. 16161780
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 16161780

Brand: Abnova™ P5242.1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for SDS-PAGE

#REF!

Specifications

Accession Number P01012
For Use With (Application) SDS-PAGE
Gene ID (Entrez) 396058
Molecular Weight (g/mol) 45kDa
Name Ovalbumin (Egg White)
Preparation Method Native protein chromatographically purified from Egg White
Quantity 1 g
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name SERPINB14
Gene Symbol SERPINB14
Assays Conjugo-immuno determinations; Drug and pharmaceutical processing.
Species Chicken
Protein Tag None
Expression System Native protein chromatographically purified from Egg White
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.