missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTP Antibody (CL11222) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21188CL1
This item is not returnable.
View return policy
Description
OTP Monoclonal antibody specifically detects OTP in Human,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| OTP | |
| Monoclonal | |
| CoraFluor 1 | |
| homeobox protein orthopedia, MGC3161, orthopedia homeobox, orthopedia homolog, orthopedia homolog (Drosophila) | |
| This antibody was developed using a recombinant protein derived from Q5XKR4, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Neuroscience | |
| 23440 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL11222 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction