missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Osteocrin Monoclonal Antibody (311413)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561189
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Hormone that acts as a regulator of dendritic growth in the developing cerebral cortex in response to sensory experience. Induced in the brain following membrane depolarization and inhibits dendritic branching in neurons of the developing cortex.
Specifications
| Osteocrin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P61364, P61366 | |
| OSTN | |
| E. coli-derived recombinant mouse Osteocrin, Val28-Gly130. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 311413 | |
| Unconjugated | |
| OSTN | |
| MUSCLIN; Ostc; osteocrin; Ostn; Processed Osteocrin | |
| Rat | |
| Protein A/G | |
| RUO | |
| 239790, 344901 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction