missing translation for 'onlineSavingsMsg'
Learn More

Orion™ 2230XP Silica Analyzer

Product Code. 17765804
Change view
Click to view available options
Detection Limits:
0.5ppb
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detection Limits Unit Size
17765804 0.5ppb Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17765804 Supplier Thermo Scientific Orion Supplier No. 2230XP
  • ONLINE EXCLUSIVE PRICE
    17661.00€
    16209.27€ / Pack of 1
    Save 1451.73€
    8% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protect critical systems and get continuous online monitoring of reactive silica with the Thermo Scientific™ Orion™ 2230XP silica analyzer. This analyzer provides online performance and is designed to lower reagent consumption and reduce the total cost of ownership.

Product Features
  • Simple user interface offers intuitive menu navigation
  • Large display can be easy to read in different lighting conditions
  • Compact size allows analyzer to be panel or wall mounted
  • Using less reagent volumes reduces operating costs
† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
TRUSTED_SUSTAINABILITY

Specifications

Accuracy Less than 5% of reading or ±0.5ppb, whichever is greater, from 0 to 300ppb. Less than 10% of reading from 300 to 5000ppb.
Communication Details RS232 ASCII protocol for data reporting
Current Loops Two 0/4-20mA – Direct or Reverse Acting (Isolated). Maximum 900ohm load.
Description 2230XP Silica Analyzer
Detection Limits 0.5ppb
Temperature (Metric) Operating 0°C to 45°C
Type Silica Analyzer
Enclosure Rating NEMA12 (Electronics cabinet), NEMA4X (Fluidics cabinet)
Flow Rate 50 to 1000mL/min
Inlet/Outlet 1/4 in. O.D. flexible tubing, polypropylene or similar material
Max. Relative Humidity 90% at 40°C
Measurement Ranges 0 to 5000ppb auto-ranging or user Programmable. Minimum Programmable Span is 0 to 100ppb.
Methods Optical absorption at 810nm
Pressure 5psi max.
Regulatory Details Safety CE: EN/IEC61010-1, cTUVus; EMC CE: EN61326; FCC Part 15 Class A
Relay Details Four SPDT, 5A at 240V - Programmable
Repeatability Less than ±2% of reading or ±0.5ppb, whichever is greater from 0 to 300ppb. Less than ±5% of reading from 300 to 5000ppb.
Resolution 0.5ppb in all ranges
Response Time Less than 15 minutes per analysis
Sample Pressure 5psi max.
Sample Streams One
Sample Supply Continuous
Suspended Solids Less than 60μm
Temperature (English) Operating 32°F to 113°F
Dimensions (L x W x H) 27 x 15 x 6 in. (695 x 395 x 158mm)
Length (English) 27 in.
Length (Metric) 695 mm
Width (English) 15 in.
Width (Metric) 395 mm
Height (English) 6 in.
Height (Metric) 158 mm
Weight (English) 40 lb.
Weight (Metric) 18 kg
Electrical Requirements 100 to 240VAC, 50/60Hz
Voltage 100/240 VAC
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.