Learn More
Orientin 100 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer
Product Code. 30322569
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322569 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30322569 Supplier Dr. Ehrenstorfer Supplier No. DREA15743000WL100
 Restricted Item
Estimated Shipment: 09-12-2026
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Orientin
CAS 28608-75-5
Analyte or Component Names Orientin
Shipping Condition +20°C
Molecular Formula C21 H20 O11
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C21H20O11/c22-6-14-17(28)18(29)19(30)21(32-14)16-11(26)4-10(25)15-12(27)5-13(31-20(15)16)7-1-2-8(23)9(24)3-7/h1-5,14,17-19,21-26,28-30H,6H2/t14-,17-,18+,19-,21+/m1/s1
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc(O)c3C(=O)C=C(Oc23)c4ccc(O)c(O)c4
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]chromen-4-one
Solvent or Matrix Acetonitrile/Water
Molecular Weight (g/mol) 448.38
Quantity 1 mL
Formula Weight 448.1006 g/mol