Learn More
Oregon Pesticide Mixture 1 600 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
Specifications
Specifications
| Chemical Name or Material | Abamectin∣Spinosad |
| CAS | 71751-41-2, 168316-95-8 |
| Analyte or Component Names | Abamectin∣Spinosad |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 5 x 1 mL |
| Concentration or Composition (by Analyte or Components) | Abamectin 600 μg/mL∣Spinosad 600 μg/mL |