missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR2S2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
196.00€ - 468.00€
Specifications
| Antigen | OR2S2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
OR2S2 Polyclonal antibody specifically detects OR2S2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| OR2S2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 56656 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| olfactory receptor 2S2, Olfactory receptor OR9-3, olfactory receptor, family 2, subfamily S, member 2, OR37A, OST715 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human OR2S2 (NP_063950.2). MEVTNVIFLGVPVLFISFSYVFIITTILRIPSAEGRKKVFSTCSAHLTVVIVFYGTLFFMYGKPKSKDSMGADKEDLSDKLIPLFYGVVTPMLNPIIYSLR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title