missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGFOD2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76533-0.025mg
This item is not returnable.
View return policy
Description
OGFOD2 Polyclonal specifically detects OGFOD2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| OGFOD2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| 2-oxoglutarate and iron-dependent oxygenase domain containing 2, DKFZp686H15154, EC 1.14.11.-, FLJ13491, FLJ37501, MGC120434, MGC120436,2-oxoglutarate and iron-dependent oxygenase domain-containing protein 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79676 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000 | |
| NP_078899 | |
| OGFOD2 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human OGFOD2. The immunogen is located within amino acids 140 - 190 of OGFOD2. Amino Acid Squence: EHFEQSDMPKGRPNTMN | |
| 0.025 mg | |
| Primary | |
| OGFOD2 antibody is predicted to not cross-react with other TPA1 protein family members. At least three isoforms are known to exist; this antibody will only detect the two larger isoforms. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction