missing translation for 'onlineSavingsMsg'
Learn More

ODR4/TTG1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

216.00€ - 326.00€

Specifications

Antigen ODR4/TTG1
Dilution Immunocytochemistry/ Immunofluorescence 1:100-1:500
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18616542
View Documents
Novus Biologicals
NBP2-98046-100ul
100 μg
326.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18689883
View Documents
Novus Biologicals
NBP2-98046-50ul
50 μg
216.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ODR4/TTG1 Polyclonal antibody specifically detects ODR4/TTG1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

ODR4/TTG1
Immunofluorescence
Unconjugated
Rabbit
Human
chromosome 1 open reading frame 27, FLJ20505, hODR-4, LAG1-interacting protein, odorant response abnormal 4, odr-4, ODR4, protein odr-4 homolog, Transactivated by transforming growth factor beta protein 1, TTG1, TTG1A
Produced in rabbits immunized with E. coli-derived Human ODR4/TTG1 fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunocytochemistry/ Immunofluorescence 1:100-1:500
Polyclonal
Purified
RUO
PBS (pH 7.0)
54953
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.