missing translation for 'onlineSavingsMsg'
Learn More

OCT4 Antibody (CL7621), Novus Biologicals™

Mouse Monoclonal Antibody

329.00€ - 600.00€

Specifications

Antigen OCT4
Clone CL7621
Dilution Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml
Applications Western Blot, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18642595
View Documents
Novus Biologicals
NBP3-21158-100ul
100 μg
600.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18630164
View Documents
Novus Biologicals
NBP3-21158-25ul
25 μg
329.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

OCT4 Monoclonal antibody specifically detects OCT4 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

OCT4
Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml
Monoclonal
Purified
RUO
Human
class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein
This antibody was developed against a Synthetic Peptide of Q01860. Actual immunogen sequence is proprietary.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
CL7621
Western Blot, Immunofluorescence
Unconjugated
Mouse
Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators
PBS, pH 7.2, 40% glycerol
5460
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.