missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Occludin Mouse anti-Human, Clone: OCLN/2181, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07137-100ug
This item is not returnable.
View return policy
Description
Occludin Monoclonal specifically detects Occludin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| Occludin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| BLCPMG, occludin | |
| Recombinant fragment of human Occludin protein (around aa 282-415) (exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| OCLN/2181 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 100506658 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction