missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCAM/NCAM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00€ - 307.65€
Specifications
| Antigen | OCAM/NCAM2 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18658242
|
Novus Biologicals
NBP2-97119-100ul |
100 μg |
326.00€ 307.65€ / 100µL Save 18.35€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653712
|
Novus Biologicals
NBP2-97119-50ul |
50 μg |
227.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OCAM/NCAM2 Polyclonal antibody specifically detects OCAM/NCAM2 in Human samples. It is validated for ELISASpecifications
| OCAM/NCAM2 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Growth and Development, Membrane Vesicle Markers, Neuronal Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 4685 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| N-CAM-2, NCAM-2, NCAM21MGC51008, neural cell adhesion molecule 2, neural cell adhesion molecule 2 precursor10 | |
| Produced in rabbits immunized with purified, recombinant Human OCAM/NCAM2 (Accession#: NP_004531.2; Met1-Asn697) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title