missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OAT1 Polyclonal antibody specifically detects OAT1 in Human, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | OAT1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | hOAT1, hPAHT, hROAT1, MGC45260, OAT1FLJ55736, Organic anion transporter 1, PAH transporter, PAHTHOAT1, para-aminohippurate transporter, Renal organic anion transporter 1, ROAT1, solute carrier family 22 (organic anion transporter), member 6, solute carrier family 22 member 6 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 31-135 of human OAT1 (NP_004781.2). MASHNTLQNFTAAIPTHHCRPPADANLSKNGGLEVWLPRDRQGQPESCLRFTSPQWGLPFLNGTEANGTGATEPCTDGWIYDNSTFPSTIVTEWDLVCSHRALRQ |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?