Learn More
pH Calibration Buffer Bottles, Oakton™
Product Code. 11750079 Shop All Cole-Parmer Products
Change view
Click to view available options
pH:
1.68
4.01
7.00
10.00
12.46
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Unit Size
11750079 10.00 Each
11739679 1.68 Each
10038434 4.01 Each
10303765 7.00 Each
11710069 12.46 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11750079 Supplier Cole Parmer Supplier No. 0065408
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Use these PH buffers for meters with 0.01 resolution. Oakton™ pH Calibration Buffer Bottles are standardized against NIST-traceable pH references within 0.01 pH at 77°F (25°C).
  • pH buffer solutions allow the user to adjust their system to read accurate measurements
  • Store buffers away from direct sunlight.
Order Info
Shipping weight: 1-1/4 lbs. (0.6kg).
Certifications
NIST traceable, Certified to NIST traceable standards.
Specifications
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 10.00
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Grade Not Available