Learn More
o-Phenylphenol Glucuronide, TRC
Product Code. 30138350
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138350 100 mg 100mg
17982555 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138350 Supplier Toronto Research Chemicals Supplier No. TRCP336125100MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 31016-74-7
Molecular Formula C18H18O7
Molecular Weight (g/mol) 346.33
Synonym [1,1'-Biphenyl]-2-yl β-D-Glucopyranosiduronic Acid
IUPAC Name 3,4,5-trihydroxy-6-(2-phenylphenoxy)oxane-2-carboxylic acid
SMILES OC1C(Oc2ccccc2c3ccccc3)OC(C(O)C1O)C(=O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C18H18O7/c19-13-14(20)16(17(22)23)25-18(15(13)21)24-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9,13-16,18-21H,(H,22,23)
Formula Weight 346.1053
Chemical Name or Material o-Phenylphenol Glucuronide