CAS RN 31016-74-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
o-Phenylphenol Glucuronide, TRC
CAS: 31016-74-7 Molecular Formula: C18H18O7 Molecular Weight (g/mol): 346.33 Synonym: [1,1'-Biphenyl]-2-yl β-D-Glucopyranosiduronic Acid IUPAC Name: 3,4,5-trihydroxy-6-(2-phenylphenoxy)oxane-2-carboxylic acid SMILES: OC1C(Oc2ccccc2c3ccccc3)OC(C(O)C1O)C(=O)O