missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Benzyl Ganciclovir N-Formamide Diacetate, TRC
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
Specifications
Specifications
| Chemical Name or Material | O-Benzyl Ganciclovir N-Formamide Diacetate |
| Analyte or Component Names | O-Benzyl Ganciclovir N-Formamide Diacetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H23 N5 O7 |
| InChI Formula | InChI=1S/C21H23N5O7/c1-14(28)30-9-17(10-31-15(2)29)33-13-26-11-22-18-19(26)24-21(23-12-27)25-20(18)32-8-16-6-4-3-5-7-16/h3-7,11-12,17H,8-10,13H2,1-2H3,(H,23,24,25,27) |
| Solution Type | Neat |
| SMILES | CC(=O)OCC(COC(=O)C)OCn1cnc2c(OCc3ccccc3)nc(NC=O)nc12 |
| IUPAC Name | [3-acetyloxy-2-[(2-formamido-6-phenylmethoxypurin-9-yl)methoxy]propyl] acetate |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 457.437 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction