Learn More
Macherey-Nagel™ Nylon Polyamide Adsorbent With Fluorescent Indicator UV254
Click to view available options
Quantity:
1000 g
Unit Size:
1kg
Description
TLC adsorbents for self-coating of glass plates or foils used in thin layer chromatography.
Specifications
Specifications
| Quantity | 1000 g |
| For Use With (Application) | TLC |
| Product Type | Polyamide Adsorbent |
| Includes | Fluorescence Indicator UV254 |
| Material | Nylon |