missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NXT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57120-25ul
This item is not returnable.
View return policy
Description
NXT1 Polyclonal specifically detects NXT1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| NXT1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| MTR2NTF2-related export protein 1, NTF2-like export factor 1, NTX2-like export factor1, NUTF-like export factor 1, P15, Protein p15 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| NXT1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SGQESLSEFFEMLPSSEFQISVVD | |
| 25 μL | |
| Signal Transduction | |
| 29107 | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur