missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUTM1 Monoclonal Antibody (1G6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA562067
This item is not returnable.
View return policy
Description
NUTM1 Monoclonal Antibody for Western Blot, IHC (P)
Plays a role in the regulation of proliferation. Regulates TERT expression by modulating SP1 binding to TERT promoter binding sites.
Specifications
| NUTM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q86Y26, Q8BHP2 | |
| NUTM1 | |
| Partial recombinant protein made to an internal portion of human NUT (between amino acids 400-700). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1G6 | |
| Unconjugated | |
| NUTM1 | |
| C15orf55; FAM22H; Nuclear protein in testis; NUT; NUT family member 1; NUT midline carcinoma family member 1; NUT midline carcinoma, family member 1; NUTM1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 213765, 256646 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur