missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT22 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67921-1-IG-20UL
This item is not returnable.
View return policy
Description
The NUDT22 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NUDT22. This antibody recognizes human, rat antigen. The NUDT22 antibody has been validated for the following applications: WB, ELISA, FC (Intra) analysis.Specifications
| NUDT22 | |
| Monoclonal | |
| 1000 μg/mL | |
| MGC13045 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| 33 kDa | |
| Liquid |
| Western Blot, ELISA, Flow Cytometry (Intracellular Staining) | |
| 3C3H9 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 84304 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction